missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Vapor Pressure | 2.6mmHg (20°C) |
Quantity | 1 kg |
Linear Formula | H3BO3 |
UN Number | NONH for all modes of transport |
Solubility Information | In accordance with ethanol |
Formula Weight | 61.83g/mol |
Percent Purity | 99.5 to 100.5% |
Grade | puriss. |
Chemical Name or Material | Boric Acid, Meets analytical specification of Ph. Eur.BP, NF |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'