missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, MP Biomedicals™

Catalog Number p-7083136
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O

Used as an astringent, antiseptic, antibacterial and antifungal agent. Boric Acid, MP Biomedicals is a preservative with a wide range of applications. It is used in buffers and for calibration of polyacrylamide gel columns for the separation of oligonucle

Boric acid has a single boron; its salt forms complex equilibria in solution, with the ion B4O7-2 being the predominant species. Manufacturing and industrial uses include: for weatherproofing wood and fireproofing fabrics; as a preservative; manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, artificial gems; in nickeling baths; cosmetics; printing and dyeing, painting; photography; for impregnating wicks; electric condensers; hardening steel. It is also used as an insecticide for cockroaches and black carpet beetles.

Quantity 100 g
Physical Form White Powder
Solubility Information Soluble in water; insoluble in alcohol; freely soluble in boiling water.
Formula Weight 61.84
Percent Purity ≥99.5%
Grade ACS Reagent
compliance-icons
Product Identifier
  • Boric Acid, ACS
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : boric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'