missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
12 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Vapor Pressure | 2.6 mmHg (20°C) |
Quantity | 100 g |
Linear Formula | H3BO3 |
UN Number | NONH for all modes of transport |
Formula Weight | 61.83g/mol |
Percent Purity | ≥99.5% |
Chemical Name or Material | Boric acid |
Safety and Handling
Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : boric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'