missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5391413
163.40 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O

High quality chemicals for laboratory and specialized industrial use

Packaging Poly Bottle
Specific Gravity 1L = 1.43kg
Quantity 500 g
Physical Form Granular
Formula Weight 61.83
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Boric Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'