missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
163.40 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.43kg |
Quantity | 500 g |
Physical Form | Granular |
Formula Weight | 61.83 |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | Boric Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'