missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, extra pure, SLR, Eur.Ph, granular, Fisher Chemical™

Brand:  Fisher Chemical B/3750/60

36.49 GBP valid until 2021-10-30
Use promo code "14806" to get your promotional price.



Code : 21

Additional Details : CAS Number : 10043-35-3 Weight : 1.00000kg


 View more versions of this product

Product Code. 10102140

  • £50.30 / 1kg

In stock, ready to ship: 7

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

White
3.8 to 4.8
H3BO3
61.83g/mol
Plastic powder jar
169°C
1kg
10043-35-3
11337
Powder Solid
2.7mbar at 20°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Boric acid

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents
Certificates