missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Extra Pure, SLR, Eur. Ph., Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Boric Acid, Extra Pure, SLR, Eur. Ph.
Specifications
pH | 3.8 to 4.8 |
Color | White |
Melting Point | 169°C |
Packaging | Plastic powder jar |
Vapor Pressure | 2.7mbar at 20°C |
Quantity | 1 kg |
Physical Form | Powder Solid |
Formula Weight | 61.83g/mol |
Safety and Handling
Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'