Learn More
Boric Acid, Extra Pure, SLR, Eur. Ph., Fisher Chemical™
Brand: Fisher Chemical B/3760/65
Code : 21
Additional Details : CAS Number : 10043-35-3 Weight : 5.00000kg
Specifications
White | |
3.8 to 4.8 | |
H3BO3 | |
61.83g/mol | |
Plastic keg | |
169°C |
5kg | |
10043-35-3 | |
11337 | |
Powder Solid | |
2.7mbar at 20°C |
Safety and Handling
- Boric acid
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.