missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Electrophoresis Grade, 99.5% min, Thermo Scientific Chemicals

Catalog Number 15488669
Click to view available options
Quantity:
500 g
1 kg
5 kg
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
pH 3.8 to 4.8
Color White
Melting Point ∼185°C (decomposition)
Density 1.435 g/mL
Odor Odorless
Quantity 500 g
Physical Form Powder
Beilstein 1697939
Merck Index 14,1336
Formula Weight 61.83
Percent Purity ≥99.5%
Grade Electrophoresis
Chemical Name or Material Boric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'