missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99.99%, (trace metal basis), extra pure, ACROS Organics™

Brand:  Acros Organics 315180025

Code : NEW

Additional Details : CAS Number : 10043-35-3 Weight : 2.50000kg

Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 13438518

  • £594.83 / 2.50kg

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For ultra trace analysis, Lewis acid
Specifications

Chemical Identifiers

10043-35-3
61.831
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
B(O)(O)O

Specifications

Boric acid
2.5kg
10043-35-3
BH3O3
MFCD00011337
15, 1339
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83
99.99%
Plastic bottle
100ppm max.
White
Extra Pure
99.99%
H3BO3
01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
B(O)(O)O
61.831
CHEBI:33118
Crystals or Crystalline Powder
Trace Metal Basis
Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol.
169.0°C
Videos
SDS
Documents