missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-3256933
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O

It is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is too employed in leather manufacture, electroplating, and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽185°C (decomposition)
Density 1.43515
Flash Point None
Assay Percent Range 99.99% (metals basis)
Quantity 500 g
Physical Form &Plus;8 Mesh, Granular
Beilstein 1697939
Solubility Information It is fairly soluble in boiling water but less in cold water. Soluble in lower alcohols,acetone and moderately soluble in pyridine.
Formula Weight 61.83
Chemical Name or Material Boric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'