missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Lewis acid
Specifications
Color | White |
Melting Point | 169.0°C |
Heavy Metals (as Pb) | 50ppm max. |
Assay Percent Range | 99.5% |
Quantity | 250 g |
Linear Formula | H3BO3 |
Physical Form | Powder |
Fieser | 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70 |
Merck Index | 15, 1339 |
Solubility Information | Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol. |
Show More |
Safety and Handling
Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'