missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Borane-triphenylphosphine complex, 97%

In hydroboration reactions

Brand:  Alfa Aesar™ 042969.06

Code : NEW

Additional Details : CAS Number : 2049-55-0 Weight : 0.00500kg Transport : UN number : 3396 Chem class : 1 Pack group : II

Quantity 5g

Product Code. 11320258

  • £37.74 / 5g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Specifications

Borane-triphenylphosphine complex
Powder
C18H18BP
276.12
UN3396
White
Unpleasant
2049-55-0
97%
(C6H5)3P·BH3
MFCD00012427
189° to 191°C
5g
Air & Moisture Sensitive
Videos
SDS