missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Borane-trimethylamine complex, 97%

Brand:  Alfa Aesar™ 042964.09

Code : NEW

Additional Details : CAS Number : 75-22-9 Weight : 0.01000kg Transport : UN number : 1325 Chem class : 1 Pack group : II

Quantity 10g

Product Code. 11360248

  • £48.97 / 10g


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Specifications

Borane-trimethylamine complex
0.81g/mL
97%
72.95
UN1325
3905060
10g
Moisture Sensitive
75-22-9
Powder
(CH3)3N·BH3
MFCD00012420
93° to 95°C
172°C
64°C (147°F)
Videos
SDS