missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borane-tetrahydrofuran complex, 1M soln. in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
100 mL
500 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Color | Colorless |
Density | 0.878 g/mL |
Flash Point | -21°C (-8°F) |
Packaging | ChemSeal™ bottles |
Linear Formula | BH3/(CH2)4O |
Quantity | 100 mL |
UN Number | UN3148 |
Beilstein | 3668402 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Miscible and reacts violently with water. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'