missing translation for 'onlineSavingsMsg'
Learn More

Borane-pyridine complex, 95%, ACROS Organics™

Brand:  Acros Organics 177011000

Code : 29

Additional Details : CAS Number : 110-51-0 Weight : 0.14200kg Transport : UN number : 2810 Chem class : 1 Pack group : II


Quantity 100mL

 View more versions of this product

Product Code. 10723013

  • £162.60 / 100mL


EU Stock 5
Add to basket


Specifications

Chemical Identifiers

Borane-pyridine complex
C5H8BN
MFCD00012435
95%
92.94
Pyridineborane

Specifications

Borane-pyridine complex
0.9200g/mL
1.5300 to 1.5320
95%
C5H8BN
MFCD00012435
08,50; 09,59; 12,65
100mL
110-51-0
Glass bottle
0.92
95%
92.94
10.0° to 11.0°C
Pyridineborane
73°C
Videos
SDS