Learn More
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 186968000
Code : 29
Additional Details : CAS Number : 13292-87-0 Weight : 0.68480kg Transport : UN number : 3399 Chem class : 4.3 Pack group : I
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Borane dimethyl sulfide complex | |
800 mL | |
(CH3)2S·BH3 | |
7°C | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
B.CSC | |
75.96 | |
75.95 | |
0.856 |
13292-87-0,108-88-3 | |
C2H9BS | |
MFCD00013189 | |
15,1336 | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
(methylsulfanyl)methane borane | |
9833925 | |
AcroSeal™ Glass bottle | |
0.8560g/mL |
Safety and Handling
- Borane-methyl sulfide complex, 2M solution in Toluene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only