missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Borane-diphenylphosphine complex, 98%

Brand:  Alfa Aesar™ H37175.06

Code : NEW

Additional Details : CAS Number : 41593-58-2 Weight : 0.00500kg Transport : UN number : 2813 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 15437747

  • £183.38 / 5g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

Borane-diphenylphosphine complex
200.02
UN2813
41593-58-2
MFCD00274254

Specifications

Borane-diphenylphosphine complex
Solid
200.02
UN2813
White
Moisture Sensitive
41593-58-2
98%
MFCD00274254
47° to 49°C
5g
Videos
SDS