missing translation for 'onlineSavingsMsg'
Learn More

Borane-diphenylphosphine Complex, 94%, Thermo Scientific Chemicals

Catalog Number p-3212470
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
41593-58-2
C12H14BP
200.03
MFCD00274254
XUFGOLYVDOBYLK-UHFFFAOYSA-N
borane-diphenylphosphine complex, diphenylphosphine borane, borane diphenylphosphine complex, diphenylphosphine-borane, diphenylphosphine trihydroboron, xufgolyvdobylk-uhfffaoysa-n, diphenylphosphine boranylidyne
11095625
boron;diphenylphosphane
B.P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
41593-58-2
C12H14BP
200.03
MFCD00274254
XUFGOLYVDOBYLK-UHFFFAOYSA-N
borane-diphenylphosphine complex, diphenylphosphine borane, borane diphenylphosphine complex, diphenylphosphine-borane, diphenylphosphine trihydroboron, xufgolyvdobylk-uhfffaoysa-n, diphenylphosphine boranylidyne
11095625
boron;diphenylphosphane
B.P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 47.0°C to 50.0°C
Color White
Elemental Analysis 92.0% min. ex C% min.
Boiling Point 148.0°C to 150.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: decomposes
Formula Weight 200.03
Percent Purity 94%
Physical Form Crystals or Powder
Chemical Name or Material Borane-diphenylphosphine complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Borane-diphenylphosphine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P402-Store in a dry place.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'