missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethylamine complex, 98+%, Thermo Scientific Chemicals

Catalog Number p-4509478
12.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
74-94-2
C2H10BN
58.92
MFCD00051068
FTDUHBOCJSQEKS-UHFFFAOYSA-N
Dimethylamineborane, DMAB
dimethylamine borane
B.CNC
This item is not returnable. View return policy
74-94-2
C2H10BN
58.92
MFCD00051068
FTDUHBOCJSQEKS-UHFFFAOYSA-N
Dimethylamineborane, DMAB
dimethylamine borane
B.CNC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 33.0°C to 37.0°C
Color White to Yellow
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CH3)2NH·BH3
Quantity 5 g
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,thf,,diglyme,benzene,toluene,chlorobenzene
Formula Weight 58.92
Percent Purity 98+%
Physical Form Flakes or Granules
Chemical Name or Material Borane-dimethylamine complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Borane-dimethylamine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'