missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethylamine complex, 97%, Thermo Scientific Chemicals

Catalog Number 11319399
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
74-94-2
C2H10BN
58.92
MFCD00051068
FTDUHBOCJSQEKS-UHFFFAOYSA-N
Dimethylamine borane
dimethylamine borane
B.CNC
This item is not returnable. View return policy
74-94-2
C2H10BN
58.92
MFCD00051068
FTDUHBOCJSQEKS-UHFFFAOYSA-N
Dimethylamine borane
dimethylamine borane
B.CNC

Borane-dimethylamine complex is used in a nickel-catalyzed reduction of electron-deficient aryl tosylates. It also serves as a reducing agent in electrolytic deposition of metals, alloys, semiconductors and insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 36°C
Color White
Density 0.69 g/mL
Boiling Point 59°C to 65°C (1 to 2 mmHg)
Quantity 25 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Soluble in methanol,water,ethanol,2-propanol and terahydrofuran.
Formula Weight 58.92
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Borane-dimethylamine complex
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'