missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borane-dimethylamine complex, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Borane-dimethylamine complex is used in a nickel-catalyzed reduction of electron-deficient aryl tosylates. It also serves as a reducing agent in electrolytic deposition of metals, alloys, semiconductors and insulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 36°C |
Color | White |
Density | 0.69 g/mL |
Boiling Point | 59°C to 65°C (1 to 2 mmHg) |
Quantity | 25 g |
UN Number | UN2925 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in methanol,water,ethanol,2-propanol and terahydrofuran. |
Formula Weight | 58.92 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'