missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Borane-dimethyl sulfide complex, nominally 5M in ether, packaged under Argon in resealable ChemSeal™ bottles

Brand:  Alfa Aesar™ 043327.AK

Code : NEW

Additional Details : CAS Number : 13292-87-0 Weight : 0.01000kg Transport : UN number : 2924 Chem class : 3 Pack group : I

Quantity 250mL

 View more versions of this product

Product Code. 11379488

  • £203.20 / 250mL


Estimated Shipment date
from Supplier 20-06-2018
Add to basket

Specifications

Chemical Identifiers

Borane-dimethyl sulfide complex
C2H9BS
75.97
UN2924
13292-87-0
(CH3)2S·BH3
MFCD00013189
3663489

Specifications

Borane-dimethyl sulfide complex
0.789g/mL
Liquid
(CH3)2S·BH3
MFCD00013189
3663489
Nominally 5M in ether
−35°C (−31°F)
Air & Moisture Sensitive
13292-87-0
ChemSeal™ bottles
C2H9BS
75.97
UN2924
Colorless
250mL
Characteristic
Videos
SDS