missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Borane-2-methylpyridine complex, 95%

Brand:  Alfa Aesar™ H66642.06

Code : NEW

Additional Details : CAS Number : 3999-38-0 Weight : 0.00500kg Transport : UN number : 2813 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 15442799

  • £36.80 / 5g


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Description

Description

Applications
Used as reactant for N-Benzyl-protection of amino acid derivatives by reductive alkylation, reductive alkoxyamination, as reducing agent for the labeling of oligosaccharides by reductive amination, synthesis of alkoxyamine derivatives, via reduction of oxime ethers, reductive amination reactions of C1-C10 aldehyde 2,4-dinitrophenylhydrazones, synthesis of various trifluoromethylated amino compounds and an alternative reagent for reductive aminations.

Solubility
Slightly sol. in water, cyclohexane, very sol. in methanol, THF, diglyme, and benzene

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Borane-2-methylpyridine complex
106.96
UN2813
3999-38-0
MFCD07784361
2-Picoline borane complex

Specifications

Borane-2-methylpyridine complex
Solid
106.96
UN2813
White
5g
Moisture Sensitive
3999-38-0
95%
MFCD07784361
44° to 50°C
2-Picoline borane complex
Slightly soluble in water, cyclohexane, very sol; in methanol, THF, diglyme, and benzene
Videos
SDS