missing translation for 'onlineSavingsMsg'
Learn More

BOC-O-Benzyl-L-serine hydroxysuccinimide ester, 97%, ACROS Organics™

Brand:  Acros Organics 440890250

Code : 29

Additional Details : CAS Number : 13650-73-2 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11955551

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

BOC-O-Benzyl-L-serine hydroxysuccinimide ester
97%
392.41
≥96.0%
C19H24N2O7
BOC-Ser(Bzl)-OSu

Specifications

BOC-O-Benzyl-L-serine hydroxysuccinimide ester
Authentic
1% max.
-14° to -18° (20°C, 589nm) (c=1 in DMF)
97%
392.41
White to off-white
13650-73-2
Glass bottle
Powder
≥96.0%
C19H24N2O7
25g
BOC-Ser(Bzl)-OSu
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BOC-O-Benzyl-L-serine hydroxysuccinimide ester

Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.

SDS