missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Serine benzyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-4228425
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
59524-02-6
C15H21NO5
295.33
BQADRZHPZVQGCW-LBPRGKRZSA-N
boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester
7018792
benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
59524-02-6
C15H21NO5
295.33
BQADRZHPZVQGCW-LBPRGKRZSA-N
boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester
7018792
benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 67°C to 72°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -14° to -18° (20°C, 589nm) (c=1, ethanol)
Formula Weight 295.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material BOC-L-Serine benzyl ester
Show More Show Less
compliance-icons
Product Identifier
  • Boc-L-Serine benzyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'