missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Methionine, +99%, Thermo Scientific Chemicals

Catalog Number p-70720
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
2488-15-5
C10H19NO4S
249.33
MFCD00065586
IMUSLIHRIYOHEV-ZETCQYMHSA-N
boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl
89857
CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
This item is not returnable. View return policy
2488-15-5
C10H19NO4S
249.33
MFCD00065586
IMUSLIHRIYOHEV-ZETCQYMHSA-N
boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl
89857
CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 47.0°C to 53.0°C
Color White
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
Packaging Glass Bottle
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Specific Rotation − 23.00
Formula Weight 249.33
Percent Purity ≥99%
Water 0.5% max. (K.F.)
Quantity 5 g
Physical Form Powder
Chemical Name or Material BOC-L-Methionine, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'