missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Methionine, +99%, ACROS Organics™

Brand:  Acros Organics 275630250

Code : 29

Additional Details : CAS Number : 2488-15-5 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10686851

  • £42.60 / 25g


Estimated Shipment
18-05-2018
Add to basket

Specifications

Chemical Identifiers

BOC-L-Methionine
2488-15-5
99+%
CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
MFCD00065586
99+%
≥99%
C10H19NO4S
249.33
N-(tert-Butoxycarbonyl)-L-methionine

Specifications

BOC-L-Methionine
2488-15-5
≥99%
Authentic
CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
MFCD00065586
N-(tert-Butoxycarbonyl)-L-methionine
0.5% max. (K.F.)
47.0° to 53.0°C
99+%
Powder
99+%
C10H19NO4S
249.33
Glass bottle
− 23.00
White to off-white
25g
Videos
SDS