missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Aspartic acid 4-benzylester, 99+%, Thermo Scientific™

Catalog Number p-3757480
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
7536-58-5
C16H21NO6
323.345
MFCD00065564
SOHLZANWVLCPHK-LBPRGKRZSA-N
boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
1581888
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
This item is not returnable. View return policy
7536-58-5
C16H21NO6
323.345
MFCD00065564
SOHLZANWVLCPHK-LBPRGKRZSA-N
boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
1581888
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
Melting Point 98°C to 102°C
Color White
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula C6H5CH2OCOCH2CH(COOH)nHCOOC(CH3)3
Packaging Glass Bottle
Specific Rotation -18.5° to -21° (20°C, 589nm) (c=2, DMF)
Formula Weight 323.34
Percent Purity ≥99.0%
Quantity 5 g
Physical Form Powder
Chemical Name or Material BOC-L-Aspartic acid 4-benzylester
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'