missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BOC Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | BOC |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
BOC Polyclonal specifically detects BOC in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BOC | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Boc homolog (mouse), brother of CDO, Protein BOC | |
BOC | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
AAH78631 | |
91653 | |
Mouse myeloma cell line NS0-derived recombinant mouse BOC Asp26-Lys508 (Gln ins) Val509-Asp846 Accession # AAH78631 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse BOC in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human BOC is observed. |
For Research Use Only