missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Boc-alpha-phosphonoglycine trimethyl ester, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
Melting Point | 67°C to 71°C |
Quantity | 1 g |
Solubility Information | Insoluble in water. |
Formula Weight | 297.24 |
Percent Purity | 95% |
Chemical Name or Material | (+/-)-Boc-alpha-phosphonoglycine trimethyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'