missing translation for 'onlineSavingsMsg'
Learn More

BOC-β-alanine, 99%, Thermo Scientific Chemicals

Catalog Number p-4228406
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
3303-84-2
C8H15NO4
189.21
MFCD00037291
WCFJUSRQHZPVKY-UHFFFAOYSA-N
boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
76809
3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NCCC(O)=O
This item is not returnable. View return policy
3303-84-2
C8H15NO4
189.21
MFCD00037291
WCFJUSRQHZPVKY-UHFFFAOYSA-N
boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
76809
3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 77.5°C to 78.5°C
Color White
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 189.21
Percent Purity 99%
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material BOC-β-Alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'