missing translation for 'onlineSavingsMsg'
Learn More

BOC-β-alanine, 99%, ACROS Organics™

Brand:  Acros Organics 437651000

Code : 29

Additional Details : CAS Number : 3303-84-2 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10595860

  • £158.20 / 100g


Estimated Shipment
13-02-2018
Add to basket

Specifications

Chemical Identifiers

BOC-β-Alanine
99%
189.21
≥98.5%
C8H15NO4

Specifications

BOC-β-Alanine
50.02 to 51.54 % C 7.29 to 7.51 % N
Glass bottle
≥98.5%
C8H15NO4
189.21
White to off-white
3303-84-2
Authentic
Crystalline Powder
99%
76° to 80°C
100g
Videos
SDS