missing translation for 'onlineSavingsMsg'
Learn More

Boat microcombustion 4mm x 4 x 15mm platinum Ogussa

Brand:  N/A 2332 3002

Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 12700229

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.