Learn More
BNTX maleate, Tocris Bioscience™
Standard δ1 selective antagonist
Brand:  Tocris (Bio-Techne) 089950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488123
  • £582.37 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Standard selective δ1 opioid receptor antagonist. Also inhibits neurogenic ion transport mediated by a putative novel opioid receptor in porcine ileal mucosa.
Specifications
Chemical Identifiers
864461-31-4
545.588
7-Benzylidenenaltrexone maleate
C31H31NO8
GKHUUKGUNNYVGB-RAJBGOKPSA-N
124081000
Specifications
BNTX maleate
>99%
7-Benzylidenenaltrexone maleate
C1CC1CN2CCC34C5C(=O)C(=CC6=CC=CC=C6)CC3(C2CC7=C4C(=C(C=C7)O)O5)O.C(=CC(=O)O)C(=O)O
545.588
50mg
864461-31-4
C31H31NO8
GKHUUKGUNNYVGB-RAJBGOKPSA-N
(4R,4aR,6E,7aR,12bR)-6-benzylidene-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;(E)-but-2-enedioic acid
124081000
545.59
Videos