missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549988
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total BNIP2 protein.
BNIP2 Polyclonal antibody specifically detects BNIP2 in Human, Mouse samples. It is validated for Western Blot.Specifications
BNIP2 | |
Polyclonal | |
Unconjugated | |
Bnip2 | |
5730523P12Rik; Bcl-2 and E1B 19 kDa interacting protein; BCL2 interacting protein 2; BCL2/adenovirus E1B 19 kDa protein-interacting protein 2; BCL2/adenovirus E1B 19 kDa-interacting protein 2 beta; BCL2/adenovirus E1B 19kDa interacting protein 2; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP2; BCL2/adenovirus E1B interacting protein 1, NIP2; BCL2/adenovirus E1B interacting protein 2; BNIP 2; BNIP2; BNIP-2; BNIP2beta; NIP2; Nip21; Nip2l | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12175, 663 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O54940, Q12982 | |
Bnip2 | |
Synthesized peptide derived from C-terminal of human BNIP2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |