missing translation for 'onlineSavingsMsg'
Learn More

BMY 7378 dihydrochloride, Tocris Bioscience™

Selective α1D antagonist

Brand:  Tocris (Bio-Techne) 100650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458273

  • £257.41 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

5-HT1A partial agonist and high affinity α1D adrenoceptor antagonist (Ki values are 2, 800 and 600 nM at cloned rat α1D, rat α1A and hamster α1B receptors respectively). Also available as part of the α1-Adrenoceptor Tocriset™ .
Specifications

Chemical Identifiers

21102-95-4
458.424
bmy 7378 dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, unii-kc07kv8t5o, kc07kv8t5o, 1,1-cyclopentanediacetimide, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-, dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, azaspiro 4.5 decane-7,9-dione dihydrochloride, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-1,1-cyclopentanediacetimide dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, bmy7378 dihydrochloride
8-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-8-azaspiro[4.5]decane-7,9-dione;dihydrochloride
C22H33Cl2N3O3
NIBOMXUDFLRHRV-UHFFFAOYSA-N
210172
COC1=CC=CC=C1N2CCN(CC2)CCN3C(=O)CC4(CCCC4)CC3=O.Cl.Cl

Specifications

BMY 7378 dihydrochloride
21102-95-4
bmy 7378 dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, unii-kc07kv8t5o, kc07kv8t5o, 1,1-cyclopentanediacetimide, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-, dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, azaspiro 4.5 decane-7,9-dione dihydrochloride, n-2-4-o-methoxyphenyl-1-piperazinyl ethyl-1,1-cyclopentanediacetimide dihydrochloride, 8-2-4-2-methoxyphenyl piperazin-1-yl ethyl-8-azaspiro 4.5 decane-7,9-dione dihydrochloride, bmy7378 dihydrochloride
COC1=CC=CC=C1N2CCN(CC2)CCN3C(=O)CC4(CCCC4)CC3=O.Cl.Cl
458.424
458.42
50mg
C22H33Cl2N3O3
NIBOMXUDFLRHRV-UHFFFAOYSA-N
8-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-8-azaspiro[4.5]decane-7,9-dione;dihydrochloride
210172
>98%
Videos