missing translation for 'onlineSavingsMsg'
Learn More

BMY 14802 hydrochloride, Tocris Bioscience™

Sigma antagonist. Antipsychotic agent

Brand:  Tocris (Bio-Techne) 144010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13459503

  • £128.25 / 10mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Potent sigma receptor antagonist (IC50 = 112 nM) with modest to weak affinity for 5-HT1A and α1 receptors. Antipsychotic following oral administration and acts via indirect modulation of central dopaminergic systems.
Specifications

Chemical Identifiers

105565-55-7
384.856
bmy 14802 hydrochloride, bmy-14802 hydrochloride, bmy 14802 hcl, hcl, 1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride, 1-piperazinebutanol, alpha-4-fluorophenyl-4-5-fluoro-2-pyrimidinyl-, monohydrochloride, 1-4-fluorophenyl-4-4-5-fluoro-2-pyrimidinyl-1-piperazinyl-1-butanol monohydrochloride, 1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride;1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride
1-(4-fluorophenyl)-4-[4-(5-fluoropyrimidin-2-yl)piperazin-1-yl]butan-1-ol;hydrochloride
C18H23ClF2N4O
NIBVEFRJDFVQLM-UHFFFAOYSA-N
3086514
C1CN(CCN1CCCC(C2=CC=C(C=C2)F)O)C3=NC=C(C=N3)F.Cl

Specifications

BMY 14802 hydrochloride
105565-55-7
bmy 14802 hydrochloride, bmy-14802 hydrochloride, bmy 14802 hcl, hcl, 1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride, 1-piperazinebutanol, alpha-4-fluorophenyl-4-5-fluoro-2-pyrimidinyl-, monohydrochloride, 1-4-fluorophenyl-4-4-5-fluoro-2-pyrimidinyl-1-piperazinyl-1-butanol monohydrochloride, 1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride;1-4-fluorophenyl-4-4-5-fluoropyrimidin-2-yl piperazin-1-yl butan-1-ol hydrochloride
C1CN(CCN1CCCC(C2=CC=C(C=C2)F)O)C3=NC=C(C=N3)F.Cl
384.856
384.86
10mg
C18H23ClF2N4O
NIBVEFRJDFVQLM-UHFFFAOYSA-N
1-(4-fluorophenyl)-4-[4-(5-fluoropyrimidin-2-yl)piperazin-1-yl]butan-1-ol;hydrochloride
3086514
>98%
Videos