Learn More
IKA™ BMT-20-G Tubes
Shop All IKA Products
Click to view available options
For Use With (Equipment):
BMT-20-G
Unit Size:
Pack of 25
Specifications
Specifications
| Product Type | Tube |
| Volume Range (Metric) | 2 to 15 mL |
| For Use With (Application) | Dry milling of dry and brittle samples (e.g. kaolin, gypsum, colored pigments, tablets), cell maceration, processing of materials mixed with fluids |
| For Use With (Equipment) | BMT-20-G |
| Packaging Quantity | 25 Pk. |