missing translation for 'onlineSavingsMsg'
Learn More

BMS 753, Tocris Bioscience™

RARα-selective agonist

Brand:  Tocris (Bio-Techne) 350550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13453094

  • £764.40 / 50mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RARα-selective agonist (Ki = 2 nM).
Specifications

Chemical Identifiers

215307-86-1
351.402
d07ytl, bms hplc, 4-1,1,3,3-tetramethyl-2-oxoindene-5-amido benzoic acid, 4-1,1,3,3-tetramethyl-2-oxoindene-5-carbonyl amino benzoic acid, 4-2,3-dihydro-1,1,3,3-tetramethyl-2-oxo-1h-inden-5-yl carbonyl amino benzoic acid
4-[(1,1,3,3-tetramethyl-2-oxoindene-5-carbonyl)amino]benzoic acid
C21H21NO4
KFBPBWUZXBYJDG-UHFFFAOYSA-N
9884820
CC1(C2=C(C=C(C=C2)C(=O)NC3=CC=C(C=C3)C(=O)O)C(C1=O)(C)C)C

Specifications

50mg
215307-86-1
d07ytl, bms hplc, 4-1,1,3,3-tetramethyl-2-oxoindene-5-amido benzoic acid, 4-1,1,3,3-tetramethyl-2-oxoindene-5-carbonyl amino benzoic acid, 4-2,3-dihydro-1,1,3,3-tetramethyl-2-oxo-1h-inden-5-yl carbonyl amino benzoic acid
CC1(C2=C(C=C(C=C2)C(=O)NC3=CC=C(C=C3)C(=O)O)C(C1=O)(C)C)C
351.402
351.4
BMS 753
C21H21NO4
KFBPBWUZXBYJDG-UHFFFAOYSA-N
4-[(1,1,3,3-tetramethyl-2-oxoindene-5-carbonyl)amino]benzoic acid
9884820
>99%
Videos
Documents