missing translation for 'onlineSavingsMsg'
Learn More

BMS 493, Tocris Bioscience™

Pan-RAR inverse agonist

Brand:  Tocris (Bio-Techne) 350950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443104

  • £764.40 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Pan-retinoic acid receptor (pan-RAR) inverse agonist. Enhances nuclear corepressor (NCoR) interaction with RARs. Binding induces analogous conformational changes in all RAR types (RARα, RARβ and RARγ).
Specifications

Chemical Identifiers

215030-90-3
404.509
4-1e-2-5,6-dihydro-5,5-dimethyl-8-2-phenylethynyl-2-naphthalenyl ethenyl benzoic acid, epothilone folate conjugate, d0n2gc, bms hplc, bms-748285 folate conjugate, bristol-myers squibb, 4-e-2-5,5-dimethyl-8-phenylethynyl-5,6-dihydro-2-napht, targeted epothilone-folate conjugate endocyte technology, cancer , bms, e-4-2-5,5-dimethyl-8-phenylethynyl-5,6-dihydronaphthalen-2-yl vinyl benzoic acid, 4-e-2-5,5-dimethyl-8-phenylethynyl-5,6-dihydronaphthalen-2-yl ethenyl benzoic acid, targeted epothilone-folate conjugate endocyte technology, cancer , bristol-myers squibb
4-[(E)-2-[5,5-dimethyl-8-(2-phenylethynyl)-6H-naphthalen-2-yl]ethenyl]benzoic acid
C29H24O2
YCADIXLLWMXYKW-CMDGGOBGSA-N
9909190
CC1(CC=C(C2=C1C=CC(=C2)C=CC3=CC=C(C=C3)C(=O)O)C#CC4=CC=CC=C4)C

Specifications

50mg
215030-90-3
4-1e-2-5,6-dihydro-5,5-dimethyl-8-2-phenylethynyl-2-naphthalenyl ethenyl benzoic acid, epothilone folate conjugate, d0n2gc, bms hplc, bms-748285 folate conjugate, bristol-myers squibb, 4-e-2-5,5-dimethyl-8-phenylethynyl-5,6-dihydro-2-napht, targeted epothilone-folate conjugate endocyte technology, cancer , bms, e-4-2-5,5-dimethyl-8-phenylethynyl-5,6-dihydronaphthalen-2-yl vinyl benzoic acid, 4-e-2-5,5-dimethyl-8-phenylethynyl-5,6-dihydronaphthalen-2-yl ethenyl benzoic acid, targeted epothilone-folate conjugate endocyte technology, cancer , bristol-myers squibb
CC1(CC=C(C2=C1C=CC(=C2)C=CC3=CC=C(C=C3)C(=O)O)C#CC4=CC=CC=C4)C
404.509
404.5
BMS 493
C29H24O2
YCADIXLLWMXYKW-CMDGGOBGSA-N
4-[(E)-2-[5,5-dimethyl-8-(2-phenylethynyl)-6H-naphthalen-2-yl]ethenyl]benzoic acid
9909190
>98%
Videos
Documents