Learn More
Tocris Bioscience™ BMS 195614
Selective RARα antagonist
Brand:  Tocris Bioscience™ 3660/50
Code : Z1
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 13433374
  • £990.78 / 50mg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Neutral retinoic acid receptor (RAR) α-selective antagonist (Ki = 2.5 nM). Displays no significant effect on nuclear receptor corepressor (NCoR) binding; moderately decreases SMRT binding to RAR. Antagonizes agonist-induced coactivator (CoA) recruitment.
Specifications
Specifications
BMS 195614
>98
50mg
BMS 195614
448.51