missing translation for 'onlineSavingsMsg'
Learn More

BMS 191011, Tocris Bioscience™

Potent BKCa (KCa1.1) channel opener

Brand:  RD Systems (Bio-Techne) 2665/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13451684

  • £189.63 / 10mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent BKCa channel opener (large-conductance Ca2+-activated potassium channel, KCa1.1). Neuroprotectant in two distinct animal models of stroke (MCAO in the SHR rat and a normotensive model of focal stroke).
Specifications

Chemical Identifiers

202821-81-6
370.712
3-5-chloro-2-hydroxybenzyl-5-4-trifluoromethyl phenyl-1,3,4-oxadiazol-2 3h-one, unii-lw7mxs978n, lw7mxs978n, 3-5-chloro-2-hydroxyphenyl methyl-5-4-trifluoromethyl phenyl-1,3,4-oxadiazol-2 3h-one, 3-5-chloro-2-hydroxybenzyl-5-4-trifluoromethyl-phenyl-1,3,4-oxadiazol-2 3h-one, 3-5-chloro-2-hydroxyphenyl methyl-5-4-trifluoromethyl phenyl-1,3,4-oxadiazol-2-one, chembl45349, 1,3,4-oxadiazol-2 3h-one, 3-5-chloro-2-hydroxyphenyl methyl-5-4-trifluoromethyl phenyl
3-[(5-chloro-2-hydroxyphenyl)methyl]-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazol-2-one
C16H10ClF3N2O3
QKOWACXSXTXRKA-UHFFFAOYSA-N
10474339
C1=CC(=CC=C1C2=NN(C(=O)O2)CC3=C(C=CC(=C3)Cl)O)C(F)(F)F

Specifications

10mg
202821-81-6
3-5-chloro-2-hydroxybenzyl-5-4-trifluoromethyl phenyl-1,3,4-oxadiazol-2 3h-one, unii-lw7mxs978n, lw7mxs978n, 3-5-chloro-2-hydroxyphenyl methyl-5-4-trifluoromethyl phenyl-1,3,4-oxadiazol-2 3h-one, 3-5-chloro-2-hydroxybenzyl-5-4-trifluoromethyl-phenyl-1,3,4-oxadiazol-2 3h-one, 3-5-chloro-2-hydroxyphenyl methyl-5-4-trifluoromethyl phenyl-1,3,4-oxadiazol-2-one, chembl45349, 1,3,4-oxadiazol-2 3h-one, 3-5-chloro-2-hydroxyphenyl methyl-5-4-trifluoromethyl phenyl
C1=CC(=CC=C1C2=NN(C(=O)O2)CC3=C(C=CC(=C3)Cl)O)C(F)(F)F
370.712
370.71
BMS 191011
C16H10ClF3N2O3
QKOWACXSXTXRKA-UHFFFAOYSA-N
3-[(5-chloro-2-hydroxyphenyl)methyl]-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazol-2-one
10474339
>99%
Videos
Documents