missing translation for 'onlineSavingsMsg'
Learn More

BMP-3 Mouse anti-Human, Clone: 203706, R&D Systems™

Mouse Monoclonal Antibody

£136.16 - £420.32

Specifications

Antigen BMP-3
Clone 203706
Immunogen E. coli-derived recombinant human BMP-3 Gln363-Arg472 Accession # P12645.2
Gene ID (Entrez) 651
Gene Accession No. P12645.2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15774644 View Documents RD Systems (Bio-Techne)
MAB1876-SP
25 ug £136.16
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-06-2020
Please to purchase this item. Need a web account? Register with us today!
13448995 View Documents RD Systems (Bio-Techne)
MAB1876
500 ug £420.32
500µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-07-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BMP-3 Monoclonal antibody MAB1876SP Specifically detects BMP-3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

BMP-3
E. coli-derived recombinant human BMP-3 Gln363-Arg472 Accession # P12645.2
P12645.2
Reconstitute at 0.5 mg/mL in sterile PBS.
Primary
Detects human BMP-3 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 40% cross-reactivity with recombinant human (rh) BMP-3b/GDF-10. In direct ELISAs and Western blots, this antibody does not cross-react with rhBMP-2, -4, -5, -6, -7, or -8.
Unconjugated
Purified
RUO
203706
651
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Monoclonal
Mouse
IgG2b
Protein A or G purified from hybridoma culture supernatant
Western Blot
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

For Research Use Only