missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish BMP-2/BMP-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | BMP-2/BMP-4 |
---|---|
Clone | 169125 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
BMP-2/BMP-4 Monoclonal specifically detects BMP-2/BMP-4 in Zebrafish samples. It is validated for Western Blot.Specifications
BMP-2/BMP-4 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Zebrafish | |
NP_571417 | |
650 | |
E. coli-derived recombinant zebrafish BMP-4 Ser288-Arg400 Accession # NP_571417 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects zebrafish BMP-2 and BMP-4 in direct ELISAs and Western blots. |
169125 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BDA2, BMP2, BMP-2, BMP2A, BMP-2A, BMP2ABone morphogenetic protein 2A, BMP2B, BMP2B1, BMP4, bone morphogenetic protein 2, MCOPS6, OFC11, SSFSC | |
BMP2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only