missing translation for 'onlineSavingsMsg'
Learn More

BML-190, Tocris Bioscience™

Potent, selective CB2 ligand. Restrictions apply. See special conditions on product page.

Brand:  Tocris (Bio-Techne) 138310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13449273

  • £110.25 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective CB2 receptor ligand (Ki values are 435 nM and > 2 μM for CB2 and CB1 respectively).
Specifications

Chemical Identifiers

2854-32-2
426.897
indomethacin morpholinylamide, 2-1-4-chlorobenzoyl-5-methoxy-2-methyl-1h-indol-3-yl-1-morpholinoethanone, chembl72631, 2-1-4-chlorobenzoyl-5-methoxy-2-methylindol-3-yl-1-morpholin-4-yl ethanone, tocris-1383, lopac-i-151, bml2-c04, indomethacin morpholinylamide, solid, indomethacin morpholinylamide bml190, n-1-4-chlorobenzoyl-5-methoxy-2-methyl-3-indoleacetyl morpholine
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]-1-morpholin-4-ylethanone
C23H23ClN2O4
BJSDNVVWJYDOLK-UHFFFAOYSA-N
2415
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)N4CCOCC4

Specifications

10mg
2854-32-2
indomethacin morpholinylamide, 2-1-4-chlorobenzoyl-5-methoxy-2-methyl-1h-indol-3-yl-1-morpholinoethanone, chembl72631, 2-1-4-chlorobenzoyl-5-methoxy-2-methylindol-3-yl-1-morpholin-4-yl ethanone, tocris-1383, lopac-i-151, bml2-c04, indomethacin morpholinylamide, solid, indomethacin morpholinylamide bml190, n-1-4-chlorobenzoyl-5-methoxy-2-methyl-3-indoleacetyl morpholine
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)N4CCOCC4
426.897
426.9
BML-190
C23H23ClN2O4
BJSDNVVWJYDOLK-UHFFFAOYSA-N
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]-1-morpholin-4-ylethanone
2415
>99%
Videos
Documents