missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Digital Thermometer

A digital thermometer for general laboratory use.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.80000kg

Product Code. 11749345

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Easy to read digital display
  • Wide temperature range from −50° to +300°C
Specifications

Specifications

Thermometer
-50°C
-50°C to +300°C
Digital
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers