missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WYPALL™ L10 EXTRA+ Wipers

Economical 1-ply wipers are stronger than conventional 1-ply towels. Kimberly-Clark™ WYPALL™ L10 EXTRA+ Wipers are designed to clean with a smear-free finish.

Brand:  Kimberly-Clark™ 7492

Code : 02

Additional Details : Weight : 6.00000kg

 View more versions of this product

Product Code. 12764376

  • £58.35 / 6 rolls


Estimated Shipment
11-05-2018
Add to basket

Description

Description

  • 1-ply single-use wipe
  • AIRFLEX™ Fabric technology
  • Stronger than traditional 1-ply wipes
  • Cleans quickly without disintegrating
  • Ideal for versatile wiping and cleaning: medium-duty cleaning tasks such as polishing glass and smooth surfaces and cleaning tools
  • Supplied as a perforated large roll
  • For use in high traffic areas.
  • Can be used in conjunction with a dispenser system to control consumption and reduce waste.
Specifications

Specifications

Blue
AQUARIUS™ Wiper dispenser
6 rolls x 400 sheets
18.5cm
38cm
L10 Wiper
38 x 18.5cm
Centrefeed Roll
1-Ply
For medium duty wiping tasks such as polishing glass, smooth surfaces and cleaning tools.
AIRFLEX™ Fabric
Videos