missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WYPALL™ L10 EXTRA Wipers, Roll Control

Provides versatility and absorbency to wipe without disintegration. Kimberly-Clark™ WYPALL™ L10 EXTRA Wiper, Roll Control is available as a centerfeed roll for use in busy areas.

Brand:  Kimberly-Clark™ 7493

Code : 02

Additional Details : Weight : 5.12548kg

 View more versions of this product

Product Code. 12754386

  • £51.50 / 6 rolls


In Stock 13
Add to basket

Description

Description

  • 1 ply single use paper wiper
  • Versatile and absorbent
  • Wipes without disintegrating.
  • Ideal for everyday wiping: light duty wiping tasks such as water spills, spray and wipe jobs, drying and cleaning hands.
  • Centerfeed roll for use in busy areas.
  • Can be used with a dispensing system to help control use and reduce wastage.
Specifications

Specifications

5.202kg (11.4444 lbs.)
380 x 185mm
Centrefeed Roll
1-Ply
Pour les opérations quotidiennes de nettoyage, les petites tâches d’essuyage fréquentes et les petites projections
AIRFLEX™ Fabric
Blue
AQUARIUS™ wiper dispenser
6 rolls x 525 sheets
1-ply
380 mm
Wypall L10 Wiper
Videos