missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Blue Graduated Class AS Clear Titration Apparatus with Lateral PTFE Valve Stopcock

Brand:  Hirschmann Instruments™ 3440370

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 11742372

  • £385.00 / Pack of 2


Estimated Shipment
24-05-2018
Add to basket

Specifications

Specifications

DIN EN ISO 385
2000mL clear glass storage bottle
Glass
Titration Apparatus
0.05mL
500mm
25mL
0.03
Videos